Assay Details
Target Gene Details
Entrez Gene ID: | 91768 |
Gene Name: | Cdk5 and Abl enzyme substrate 1 |
Gene Aliases: |
CABL1, CABLES, HsT2563, IK3-1 |
Location: |
Chr.18:23134564-23260470 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CABLES1 | NM_001100619.2 | 1 | 412 | NP_001094089.1 |
| NM_001256438.1 | NP_001243367.1 | |||
| NR_023359.1 | ||||
| AK093243.1 | BAC04107.1 | |||
| AK296407.1 | ||||
| EF028204.1 | 1 | 412 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1071838 | Chr.18:23135335 - 23136236 on Build GRCh38 | Deletion |
|
| esv27368 | Chr.18:23135613 - 23136324 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map