Assay Details
Target Gene Details
Entrez Gene ID: | 152189 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 8 |
Gene Aliases: |
CKLFSF8, CKLFSF8-V2 |
Location: |
Chr.3:32238679-32370325 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTM8 | NM_001320308.1 | NP_001307237.1 | ||
| NM_178868.4 | NP_849199.2 | |||
| XM_011533416.2 | XP_011531718.1 | |||
| XM_017005779.1 | XP_016861268.1 | |||
| AF474370.2 | AAN73034.1 | |||
| AY598783.1 | ||||
| BC041390.2 | AAH41390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3385139 | Chr.3:32297912 - 32297931 on Build GRCh38 | Insertion |
|
| nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
|
| nsv590046 | Chr.3:32297503 - 32302814 on Build GRCh38 | Loss |
|
| esv2676585 | Chr.3:32297446 - 32298338 on Build GRCh38 | Deletion |
|
| esv23045 | Chr.3:32297501 - 32298032 on Build GRCh38 | Loss |
|
| esv3595770 | Chr.3:32297309 - 32298230 on Build GRCh38 | Loss |
|
| esv3307004 | Chr.3:32297906 - 32297907 on Build GRCh38 | Mobile Element Insertion |
|
More Information
Set Membership: |
|

Genomic Map