Assay Details
Target Gene Details
Entrez Gene ID: | 9372 |
Gene Name: | zinc finger FYVE-type containing 9 |
Gene Aliases: |
MADHIP, NSP, PPP1R173, SARA, SMADIP |
Location: |
Chr.1:52142001-52348664 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFYVE9 | NM_004799.3 | NP_004790.2 | ||
| NM_007324.3 | NP_015563.2 | |||
| XM_017002845.1 | XP_016858334.1 | |||
| AF130419.1 | AAD31694.1 | |||
| AF130420.1 | AAD31695.1 | |||
| DR005493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv273n54 | Chr.1:52137527 - 52144342 on Build GRCh38 | Gain |
|
| nsv546232 | Chr.1:52141905 - 52142493 on Build GRCh38 | Loss |
|
| esv25536 | Chr.1:52141877 - 52142547 on Build GRCh38 | Loss |
|
| nsv546234 | Chr.1:52141979 - 52142493 on Build GRCh38 | Gain |
|
| nsv546237 | Chr.1:52142030 - 52151497 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map