Assay Details
Target Gene Details
Entrez Gene ID: | 100652833 |
Gene Name: | putative POM121-like protein 1-like |
Gene Aliases: |
- |
Location: |
Chr.5:99522217-99526100 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100652833 | XM_011543783.2 | 1 | 3369 | XP_011542085.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27807 | Chr.5:99517800 - 99536729 on Build GRCh38 | Gain+Loss |
|
| nsv1022266 | Chr.5:99326944 - 99732957 on Build GRCh38 | Gain |
|
| nsv964912 | Chr.5:99521566 - 99536611 on Build GRCh38 | Duplication |
|
| nsv599118 | Chr.5:99022487 - 99693749 on Build GRCh38 | Loss |
|
| esv3353357 | Chr.5:99522049 - 99524247 on Build GRCh38 | Duplication |
|
| nsv1023117 | Chr.5:99417695 - 99599530 on Build GRCh38 | Gain |
|
| esv2759358 | Chr.5:99269621 - 99553890 on Build GRCh38 | Gain |
|
| nsv10726 | Chr.5:99518205 - 99537200 on Build GRCh38 | Loss |
|
| esv3575864 | Chr.5:99403860 - 99574143 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145676920] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map