Assay Details
Target Gene Details
Entrez Gene ID: | 121022 |
Gene Name: | mitochondrial ribosomal protein S6 |
Gene Aliases: |
AW046321 |
Location: |
Chr.16:92058336-92112227 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps6 | NM_080456.1 | 3 | 558 | NP_536704.1 |
| AB049943.1 | BAB40996.1 | |||
| AK165590.1 | 3 | 575 | BAE38278.1 | |
| AK166374.1 | 3 | 573 | BAE38738.1 | |
| AK167304.1 | 3 | 573 | BAE39408.1 | |
| AK167316.1 | 3 | 573 | BAE39418.1 | |
| AY061856.1 | 3 | 573 | AAL35740.1 | |
| BC027271.1 | 3 | 558 | AAH27271.1 |
More Information
Set Membership: |
|

Genomic Map