Assay Details
Target Gene Details
Entrez Gene ID: | 15976 |
Gene Name: | interferon (alpha and beta) receptor 2 |
Gene Aliases: |
AI747302, Ifnar-2 |
Location: |
Chr.16:91372215-91405589 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifnar2 | NM_001110498.1 | 5 | 638 | NP_001103968.1 |
| NM_010509.2 | 5 | 638 | NP_034639.2 | |
| AF013274.1 | 5 | 416 | AAC53351.1 | |
| AF013486.1 | 5 | 510 | AAC53352.1 | |
| AK003185.1 | 5 | 572 | BAB22628.1 | |
| AK010952.1 | 5 | 589 | BAB27287.3 | |
| AK139169.1 | 5 | 618 | BAE23909.1 | |
| AK150386.1 | 5 | 572 | BAE29516.1 | |
| AK150597.1 | 5 | 551 | BAE29690.1 | |
| AK151474.1 | 5 | 566 | BAE30430.1 | |
| AK151493.1 | 5 | 551 | BAE30446.1 | |
| AK151777.1 | 5 | 566 | ||
| AK152878.1 | 5 | 572 | BAE31563.1 | |
| AK153317.1 | 5 | 572 | BAE31897.1 | |
| AK199902.1 | ||||
| AK209089.1 | ||||
| BC003743.1 | 4 | 333 | AAH03743.1 | |
| BC071225.1 | 4 | 336 | AAH71225.1 | |
| Y09813.1 | 5 | 393 | CAA70943.1 | |
| Y09864.1 | 5 | 528 | CAA70991.1 | |
| Y09865.1 | 5 | 528 | CAA70992.1 |
More Information
Set Membership: |
|

Genomic Map