Assay Details
Target Gene Details
Entrez Gene ID: | 17858 |
Gene Name: | MX dynamin-like GTPase 2 |
Gene Aliases: |
AI528743, Mx-2 |
Location: |
Chr.16:97536081-97560901 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mx2 | NM_013606.1 | 13 | 1778 | NP_038634.1 |
| NR_003508.1 | 13 | 1781 | ||
| AB029920.1 | 13 | 1778 | BAA82593.1 | |
| AI528743.1 | ||||
| AK143831.1 | 13 | 1782 | ||
| BC007127.1 | 13 | 1763 | AAH07127.1 | |
| J03368.1 | 13 | 1779 | AAA39778.1 |
More Information
Set Membership: |
|

Genomic Map