Assay Details
Target Gene Details
Entrez Gene ID: | 22129 |
Gene Name: | tetratricopeptide repeat domain 3 |
Gene Aliases: |
2610202A04Rik, AA409221, D16Ium21, D16Ium21e, TPRD, mKIAA4119 |
Location: |
Chr.16:94370739-94469222 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc3 | NM_009441.2 | NP_033467.2 | ||
| AB008516.1 | BAA31563.1 | |||
| AK053765.1 | BAC35513.1 | |||
| AK084201.1 | ||||
| AK088273.1 | BAC40250.1 | |||
| AK089974.1 | BAC41023.1 | |||
| AK165850.1 | BAE38412.1 | |||
| AK166120.1 | ||||
| AK166164.1 | ||||
| AK166298.1 | ||||
| AK170087.1 | ||||
| AK220495.1 | BAD90293.1 | |||
| BC019173.1 | ||||
| BC049277.1 | ||||
| BC057207.1 | AAH57207.1 |
More Information
Set Membership: |
|

Genomic Map