Assay Details
Target Gene Details
Entrez Gene ID: | 13508 |
Gene Name: | Down syndrome cell adhesion molecule |
Gene Aliases: |
4932410A21Rik |
Location: |
Chr.16:96587452-97170735 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dscam | NM_031174.4 | 31 | 5778 | NP_112451.1 |
| AF315558.1 | 31 | 5423 | AAG28796.1 | |
| AK134979.1 | 3 | 331 | ||
| AK141963.1 | 20 | 2845 | ||
| AY005483.1 | 31 | 5778 | AAF99440.1 |
More Information
Set Membership: |
|

Genomic Map