Assay Details
Target Gene Details
Entrez Gene ID: | 66609 |
Gene Name: | crystallin, zeta (quinone reductase)-like 1 |
Gene Aliases: |
2210407J23Rik, 2410006O11Rik, QOH-1 |
Location: |
Chr.16:91689322-91728802 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cryzl1 | NM_026994.1 | 9 | 959 | NP_081270.1 |
| NM_133679.2 | 10 | 1004 | NP_598440.1 | |
| AK008846.1 | 10 | 777 | ||
| AK017995.1 | 9 | 959 | ||
| AK051915.1 | 1 | 2431 | ||
| AK145827.1 | 10 | 828 | BAE26679.1 | |
| AK170997.1 | 2 | 120 | ||
| BC010479.1 | 10 | 801 | AAH10479.1 | |
| BC019387.1 | 9 | 768 | AAH19387.1 | |
| BY717916.1 |
Target Gene Details
Entrez Gene ID: | 100038392 |
Gene Name: | predicted gene 10785 |
Gene Aliases: |
ENSMUSG00000074911 |
Location: |
Chr.16:91688898-91715755 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10785 | NR_040389.1 | |||
| AK038992.1 | ||||
| AK160609.1 | BAE35912.1 |
More Information
Set Membership: |
|

Genomic Map