Assay Details
Target Gene Details
Entrez Gene ID: | 71458 |
Gene Name: | BCL6 interacting corepressor |
Gene Aliases: |
5830466J11Rik, 8430401K06Rik, BcorR, D930024N20Rik, mKIAA1575 |
Location: |
Chr.X:12036737-12160355 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcor | NM_001168321.1 | 9 | 4320 | NP_001161793.1 |
| NM_029510.3 | 10 | 5145 | NP_083786.2 | |
| NM_175044.3 | 9 | 5091 | NP_778209.2 | |
| NM_175045.3 | 10 | 5043 | NP_778210.2 | |
| NM_175046.3 | 9 | 4989 | NP_778211.2 | |
| AK018370.1 | 2 | 400 | ||
| AK031282.1 | 2 | 448 | ||
| AK129398.1 | 9 | 4320 | BAC98208.1 | |
| AY161170.1 | 10 | 4460 | AAN85318.1 | |
| AY161171.1 | 9 | 4406 | AAN85319.1 | |
| AY161172.1 | 10 | 4358 | AAN85320.1 | |
| AY161173.1 | 9 | 4304 | AAN85321.1 | |
| BC058656.1 | 10 | 4544 | AAH58656.1 |

Genomic Map