Assay Details
Target Gene Details
Entrez Gene ID: | 71458 |
Gene Name: | BCL6 interacting corepressor |
Gene Aliases: |
5830466J11Rik, 8430401K06Rik, BcorR, D930024N20Rik, mKIAA1575 |
Location: |
Chr.X:12036737-12160355 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcor | NM_001168321.1 | 6 | 3494 | NP_001161793.1 |
| NM_029510.3 | 7 | 4217 | NP_083786.2 | |
| NM_175044.3 | 6 | 4163 | NP_778209.2 | |
| NM_175045.3 | 7 | 4217 | NP_778210.2 | |
| NM_175046.3 | 6 | 4163 | NP_778211.2 | |
| AK129398.1 | 6 | 3494 | BAC98208.1 | |
| AY161170.1 | 7 | 3532 | AAN85318.1 | |
| AY161171.1 | 6 | 3478 | AAN85319.1 | |
| AY161172.1 | 7 | 3532 | AAN85320.1 | |
| AY161173.1 | 6 | 3478 | AAN85321.1 | |
| BC058656.1 | 7 | 3718 | AAH58656.1 | |
| CD563463.1 |

Genomic Map