Assay Details
Target Gene Details
Entrez Gene ID: | 70495 |
Gene Name: | ATPase, H+ transporting, lysosomal accessory protein 2 |
Gene Aliases: |
(P)RR, 5730403E06Rik, APT6M8-9, ATP6M8-9, Atp6ip2, M8-9 |
Location: |
Chr.X:12587759-12617051 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6ap2 | NM_027439.4 | NP_081715.1 | ||
| AB192471.2 | ||||
| AK017482.1 | BAB30766.2 | |||
| AK029405.1 | BAC26436.1 | |||
| AK076495.1 | BAC36365.1 | |||
| AK151735.1 | ||||
| AK168139.1 | ||||
| BC014706.1 | AAH14706.1 |
More Information
Set Membership: |
|

Genomic Map