Assay Details
Target Gene Details
Entrez Gene ID: | 71458 |
Gene Name: | BCL6 interacting corepressor |
Gene Aliases: |
5830466J11Rik, 8430401K06Rik, BcorR, D930024N20Rik, mKIAA1575 |
Location: |
Chr.X:12036737-12160355 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcor | NM_001168321.1 | 14 | 5673 | NP_001161793.1 |
| NM_029510.3 | 15 | 6498 | NP_083786.2 | |
| NM_175044.3 | 14 | 6444 | NP_778209.2 | |
| NM_175045.3 | 15 | 6396 | NP_778210.2 | |
| NM_175046.3 | 14 | 6342 | NP_778211.2 | |
| AK018034.1 | 5 | 1263 | ||
| AK031282.1 | 7 | 1800 | ||
| AK129398.1 | BAC98208.1 | |||
| BC058656.1 | 15 | 5897 | AAH58656.1 |

Genomic Map