Assay Details
Target Gene Details
Entrez Gene ID: | 17857 |
Gene Name: | MX dynamin-like GTPase 1 |
Gene Aliases: |
AI893580, Mx, Mx-1 |
Location: |
Chr.16:97447035-97462906 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mx1 | NM_010846.1 | 9 | 1784 | NP_034976.1 |
| NR_003520.1 | 8 | 1265 | ||
| AK076430.1 | 13 | 1536 | BAC36340.1 | |
| AK150023.1 | 8 | 1264 | BAE29248.1 | |
| AK150236.1 | 8 | 1264 | BAE29400.1 | |
| BC011113.1 | 8 | 1769 | AAH11113.1 | |
| EU638327.1 | 4 | 589 | ||
| EU638328.1 | 7 | 1082 | ||
| M12279.1 | 9 | 1784 | AAA39776.1 | |
| M21038.1 | 10 | 1856 | ||
| M21039.1 | 10 | 1360 |
More Information
Set Membership: |
|

Genomic Map