Assay Details
Target Gene Details
Entrez Gene ID: | 11820 |
Gene Name: | amyloid beta (A4) precursor protein |
Gene Aliases: |
Abeta, Abpp, Adap, Ag, Cvap, E030013M08Rik, betaApp |
Location: |
Chr.16:84954436-85173952 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| App | NM_001198823.1 | 18 | 2504 | NP_001185752.1 |
| NM_001198824.1 | 17 | 2447 | NP_001185753.1 | |
| NM_001198825.1 | 16 | 2393 | NP_001185754.1 | |
| NM_001198826.1 | 17 | 2450 | NP_001185755.1 | |
| NM_007471.3 | 16 | 2279 | NP_031497.2 | |
| AK052448.1 | 6 | 700 | BAC34997.1 | |
| AK076506.1 | 10 | 1199 | BAC36369.1 | |
| AK128971.1 | 16 | 2402 | ||
| AK133639.1 | 17 | 2444 | ||
| AK147543.1 | 16 | 2315 | BAE27986.1 | |
| AK148439.1 | 17 | 2323 | ||
| AK159245.1 | 17 | 2459 | BAE34927.1 | |
| AK159718.1 | 17 | 2436 | BAE35313.1 | |
| AK161854.1 | 11 | 1360 | ||
| AK161976.1 | 16 | 2254 | ||
| AY267348.1 | AAP23169.1 | |||
| BC005490.1 | 14 | 1868 | AAH05490.1 | |
| BC070409.1 | 16 | 2275 | AAH70409.1 | |
| M18373.1 | 16 | 2268 | AAA37139.1 | |
| U84012.1 | 16 | 2268 | AAB41502.1 | |
| X59379.1 | 16 | 2198 |
More Information
Set Membership: |
|

Genomic Map