Assay Details
Target Gene Details
Entrez Gene ID: | 11820 |
Gene Name: | amyloid beta (A4) precursor protein |
Gene Aliases: |
Abeta, Abpp, Adap, Ag, Cvap, E030013M08Rik, betaApp |
Location: |
Chr.16:84954436-85173952 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Target Gene Details
Entrez Gene ID: | 100038614 |
Gene Name: | predicted gene 10791 |
Gene Aliases: |
ENSMUSG00000074933 |
Location: |
Chr.16:84972211-84979451 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10791 | NR_045889.1 | |||
| AK082307.1 | 3 | 2201 | ||
| AK166404.1 | BAE38756.1 |
More Information
Set Membership: |
|

Genomic Map