Assay Details
Target Gene Details
Entrez Gene ID: | 11820 |
Gene Name: | amyloid beta (A4) precursor protein |
Gene Aliases: |
Abeta, Abpp, Adap, Ag, Cvap, E030013M08Rik, betaApp |
Location: |
Chr.16:84954436-85173952 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
More Information
Set Membership: |
|

Genomic Map