Assay Details
Target Gene Details
Entrez Gene ID: | 11820 |
Gene Name: | amyloid beta (A4) precursor protein |
Gene Aliases: |
Abeta, Abpp, Adap, Ag, Cvap, E030013M08Rik, betaApp |
Location: |
Chr.16:84954436-85173952 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| App | NM_001198823.1 | NP_001185752.1 | ||
| NM_001198824.1 | NP_001185753.1 | |||
| NM_001198825.1 | NP_001185754.1 | |||
| NM_001198826.1 | NP_001185755.1 | |||
| NM_007471.3 | NP_031497.2 | |||
| AK128971.1 | ||||
| AK133639.1 | ||||
| AK147543.1 | BAE27986.1 | |||
| AK148439.1 | ||||
| AK159245.1 | BAE34927.1 | |||
| AK159352.1 | ||||
| AK159718.1 | BAE35313.1 | |||
| AK161854.1 | ||||
| AK161976.1 | ||||
| AK193254.1 | ||||
| AK201970.1 | ||||
| AY267348.1 | AAP23169.1 | |||
| BC005490.1 | AAH05490.1 | |||
| BC070409.1 | AAH70409.1 | |||
| M18373.1 | AAA37139.1 | |||
| M24397.1 | AAA39929.1 | |||
| U84012.1 | AAB41502.1 | |||
| X59379.1 |
More Information
Set Membership: |
|

Genomic Map