Assay Details
Target Gene Details
Entrez Gene ID: | 17857 |
Gene Name: | MX dynamin-like GTPase 1 |
Gene Aliases: |
AI893580, Mx, Mx-1 |
Location: |
Chr.16:97447035-97462906 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mx1 | NM_010846.1 | NP_034976.1 | ||
| NR_003520.1 | ||||
| AK076430.1 | BAC36340.1 | |||
| AK149635.1 | 1 | 1984 | ||
| AK149792.1 | 1 | 1984 | ||
| AK150023.1 | BAE29248.1 | |||
| AK150236.1 | BAE29400.1 | |||
| BC011113.1 | AAH11113.1 | |||
| M12279.1 | AAA39776.1 | |||
| M21038.1 | ||||
| M21039.1 |
More Information
Set Membership: |
|

Genomic Map