Assay Details
Target Gene Details
Entrez Gene ID: | 76781 |
Gene Name: | methyltransferase like 4 |
Gene Aliases: |
2410198H06Rik, A730091E08Rik, AV296509, HsT661 |
Location: |
Chr.17:94722658-94750126 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl4 | NM_176917.4 | NP_795891.2 | ||
| AK157268.1 | BAE34021.1 | |||
| BC099943.1 |
More Information
Set Membership: |
|

Genomic Map