Assay Details
Target Gene Details
Entrez Gene ID: | 225055 |
Gene Name: | F-box protein 11 |
Gene Aliases: |
C80048, Jf |
Location: |
Chr.17:87990859-88065285 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxo11 | NM_001081034.1 | 8 | 1028 | NP_001074503.1 |
| BC117884.1 | 8 | 774 | ||
| BC117885.1 | 8 | 778 | ||
| BC128479.1 | 8 | 778 |
More Information
Set Membership: |
|

Genomic Map