Assay Details
Target Gene Details
Entrez Gene ID: | 67198 |
Gene Name: | spermatogenesis associated, serine-rich 2-like |
Gene Aliases: |
2810022L02Rik, A230104H11Rik, AA987173, AI480531, AI842453, AW413586 |
Location: |
Chr.1:57774162-57948398 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spats2l | NM_144882.4 | NP_659131.2 | ||
| AK012791.1 | ||||
| AK019292.1 | ||||
| AK020712.1 | ||||
| AK031266.1 | BAC27328.1 | |||
| AK032169.1 | BAC27738.1 | |||
| AK161988.1 | ||||
| BC021879.1 | AAH21879.1 | |||
| BY317793.1 |
More Information
Set Membership: |
|

Genomic Map