Assay Details
Target Gene Details
Entrez Gene ID: | 72481 |
Gene Name: | RIKEN cDNA 2610203C22 gene |
Gene Aliases: |
2310007M24Rik |
Location: |
Chr.1:9560832-9631092 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610203C22Rik | NR_015470.1 | |||
| AK009214.1 | ||||
| AK011866.1 | ||||
| AK082741.1 |
Target Gene Details
Entrez Gene ID: | 76982 |
Gene Name: | RIKEN cDNA 3110035E14 gene |
Gene Aliases: |
AI415019, AV164974, R75066 |
Location: |
Chr.1:9601267-9627142 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110035E14Rik | NM_178399.4 | NP_848486.1 | ||
| AK043576.1 | BAC31586.1 | |||
| AK043759.1 | BAC31645.1 | |||
| BC066997.1 | AAH66997.1 |
More Information
Set Membership: |
|

Genomic Map