Assay Details
Target Gene Details
Entrez Gene ID: | 11761 |
Gene Name: | aldehyde oxidase 1 |
Gene Aliases: |
AI196512, AI255253, Ao, Aox-1, Aox-2, Aox2, Moro, Ro |
Location: |
Chr.1:58029809-58106410 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aox1 | NM_009676.2 | NP_033806.2 | ||
| AB017482.1 | BAA36834.1 | |||
| AF076216.1 | AAC99382.1 | |||
| AK137697.1 | BAE23466.1 | |||
| BC026132.1 | AAH26132.1 |
More Information
Set Membership: |
|

Genomic Map