Assay Details
Target Gene Details
Entrez Gene ID: | 66615 |
Gene Name: | autophagy related 4B, cysteine peptidase |
Gene Aliases: |
2510009N07Rik, AW048066, Apg4b, Atg4bl, Autl1 |
Location: |
Chr.1:93755033-93789529 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atg4b | NM_174874.3 | NP_777363.1 | ||
| AJ504653.1 | CAD43220.1 | |||
| AK010949.1 | ||||
| AK028712.1 | BAC26079.1 | |||
| AK031570.1 | BAC27455.1 | |||
| AK075796.1 | BAC35965.1 | |||
| AK088811.1 | BAC40587.1 | |||
| AK129242.1 | BAC98052.1 | |||
| AK144239.1 | BAE25791.1 | |||
| AK188147.1 | ||||
| BC027184.1 | AAH27184.1 |
More Information
Set Membership: |
|

Genomic Map