Assay Details
Target Gene Details
Entrez Gene ID: | 20297 |
Gene Name: | chemokine (C-C motif) ligand 20 |
Gene Aliases: |
CKb4, LARC, MIP-3A, MIP-3[a], MIP3A, ST38, Scya20, exodus-1 |
Location: |
Chr.1:83116744-83119167 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccl20 | NM_001159738.1 | NP_001153210.1 | ||
| NM_016960.2 | NP_058656.1 | |||
| AB015136.1 | BAA76955.1 | |||
| AF053313.1 | AAC78295.1 | |||
| AF099052.1 | AAC78680.1 | |||
| AJ222694.1 | CAA10948.1 | |||
| AK079862.1 | 3 | 331 | ||
| BC028504.1 | AAH28504.1 | |||
| BX525886.1 | ||||
| BY026108.1 |
More Information
Set Membership: |
|

Genomic Map