Assay Details
Target Gene Details
Entrez Gene ID: | 226751 |
Gene Name: | CDC42 binding protein kinase alpha |
Gene Aliases: |
A930014J19Rik, DMPK-like |
Location: |
Chr.1:179960093-180165606 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 20 - Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42bpa | NM_001033285.1 | NP_001028457.1 | ||
| AF492452.1 | AAM10976.1 | |||
| AK044466.1 | 2 | 245 | ||
| AK047837.1 | ||||
| BC158017.1 | ||||
| BC158095.1 |
More Information
Set Membership: |
|

Genomic Map