Assay Details
Target Gene Details
Entrez Gene ID: | 12958 |
Gene Name: | crystallin, beta A2 |
Gene Aliases: |
E130107M19Rik |
Location: |
Chr.1:74889934-74898843 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cryba2 |

Genomic Map