Assay Details
Target Gene Details
Entrez Gene ID: | 72017 |
Gene Name: | cytochrome b5 reductase 1 |
Gene Aliases: |
1500005G05Rik, B5R.1, C80155, Nqo3a2 |
Location: |
Chr.1:134405561-134411740 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyb5r1 | NM_028057.2 | NP_082333.1 | ||
| AK005159.1 | BAB23850.1 | |||
| AK049717.1 | 5 | 826 | ||
| AK159663.1 | BAE35270.1 | |||
| AK167178.1 | BAE39313.1 | |||
| AK212943.1 | ||||
| BC016266.1 | AAH16266.1 | |||
| BC024618.1 | AAH24618.1 |
More Information
Set Membership: |
|

Genomic Map