Assay Details
Target Gene Details
Entrez Gene ID: | 52392 |
Gene Name: | DNA segment, Chr 1, ERATO Doi 622, expressed |
Gene Aliases: |
AI987691, AW047450 |
Location: |
Chr.1:97643902-97662018 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D1Ertd622e | NM_133825.3 | NP_598586.1 | ||
| AK135656.1 | BAE22601.1 | |||
| AK137265.1 | BAE23288.1 | |||
| AK152370.1 | BAE31160.1 | |||
| AK162223.1 | BAE36800.1 | |||
| AK168549.1 | BAE40424.1 | |||
| BB611324.1 | ||||
| BC019375.1 | AAH19375.1 | |||
| BC023951.1 | AAH23951.1 |

Genomic Map