Assay Details
Target Gene Details
Entrez Gene ID: | 23836 |
Gene Name: | cadherin 20 |
Gene Aliases: |
Cdh7 |
Location: |
Chr.1:104768819-104995481 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh20 | NM_011800.4 | NP_035930.1 | ||
| AF007116.1 | 1 | 290 | AAD01278.1 | |
| AK034475.1 | BAC28721.1 | |||
| BC119606.2 | ||||
| BC119607.2 |
More Information
Set Membership: |
|

Genomic Map