Assay Details
Target Gene Details
Entrez Gene ID: | 14048 |
Gene Name: | eyes absent 1 homolog (Drosophila) |
Gene Aliases: |
bor |
Location: |
Chr.1:14168953-14310265 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eya1 | NM_001252192.1 | NP_001239121.1 | ||
| NM_010164.2 | NP_034294.2 | |||
| AK042412.1 | BAC31251.1 | |||
| AK045320.1 | ||||
| AK153788.1 | ||||
| BC060260.1 | AAH60260.1 | |||
| BC066860.1 | ||||
| U61110.1 | 1 | 46 | AAB48017.1 |

Genomic Map