Assay Details
Target Gene Details
Entrez Gene ID: | 214469 |
Gene Name: | family with sequence similarity 168, member B |
Gene Aliases: |
mKIAA4042 |
Location: |
Chr.1:34813218-34843050 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam168b | NM_001160235.1 | NP_001153707.1 | ||
| NM_001160236.1 | NP_001153708.1 | |||
| NM_174997.2 | NP_778162.1 | |||
| AK144298.1 | BAE25820.1 | |||
| AK220453.1 | BAD90278.1 | |||
| BC040511.1 | AAH40511.1 | |||
| BC043098.1 | AAH43098.1 | |||
| BC059247.1 | AAH59247.1 | |||
| CA328263.1 | ||||
| CF538379.1 |
More Information
Set Membership: |
|

Genomic Map