Assay Details
Target Gene Details
Entrez Gene ID: | 227058 |
Gene Name: | dynein, axonemal, heavy chain 7B |
Gene Aliases: |
Dnahc7b, Gm107 |
Location: |
Chr.1:46065786-46373551 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnah7b | NM_001160386.1 | NP_001153858.1 |
More Information
Set Membership: |
|

Genomic Map