Assay Details
Target Gene Details
Entrez Gene ID: | 226866 |
Gene Name: | somatomedin B and thrombospondin, type 1 domain containing |
Gene Aliases: |
Gm106, Rpesp |
Location: |
Chr.1:15853862-15892722 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sbspon | NM_001033288.3 | NP_001028460.1 | ||
| AK137144.1 | BAE23250.1 | |||
| AK143251.1 | BAE25326.1 | |||
| BC147194.1 | ||||
| BC147195.1 |

Genomic Map