Assay Details
Target Gene Details
Entrez Gene ID: | 240832 |
Gene Name: | torsin A interacting protein 2 |
Gene Aliases: |
1110020D10Rik, 15kDa, A130072J07, AA103493, AW060462, AW610675, C77739, Ifrg15, Lull1 |
Location: |
Chr.1:156035664-156068861 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor1aip2 | NM_001160180.1 | NP_001153652.1 | ||
| NM_001160181.1 | NP_001153653.1 | |||
| NM_001160182.1 | NP_001153654.1 | |||
| NM_022329.4 | NP_071724.1 | |||
| NM_172843.4 | NP_766431.3 | |||
| AA030492.1 | ||||
| AJ251363.1 | CAC13975.1 | |||
| AK003852.1 | BAB23037.1 | |||
| AK032697.1 | BAC27991.1 | |||
| AK038024.1 | BAC29923.1 | |||
| AK080959.1 | BAC38096.1 | |||
| AK150117.1 | BAE29320.1 | |||
| AK150303.1 | BAE29454.1 | |||
| AK151453.1 | BAE30413.1 | |||
| AK152137.1 | BAE30976.1 | |||
| AK177027.1 | ||||
| AK182358.1 | ||||
| AK203799.1 | ||||
| AK212068.1 | ||||
| AK212760.1 | ||||
| AK214292.1 | ||||
| AK216254.1 | ||||
| AK218632.1 | ||||
| AK219881.1 | ||||
| BC064017.1 | AAH64017.1 | |||
| BC092059.1 | AAH92059.1 | |||
| BU614106.1 | ||||
| BY029314.1 | ||||
| BY254976.1 | ||||
| CF164425.1 |

Genomic Map