Assay Details
Target Gene Details
Entrez Gene ID: | 329152 |
Gene Name: | HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2 |
Gene Aliases: |
A730039N16Rik, D030049F17Rik, Gm971, Nedl2, mKIAA1301 |
Location: |
Chr.1:53806872-54195034 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hecw2 | NM_001001883.3 | NP_001001883.1 | ||
| NM_172655.3 | NP_766243.2 | |||
| AB182244.1 | BAD23960.1 | |||
| AK042922.1 | BAC31406.1 | |||
| AK129325.1 | BAC98135.1 | |||
| BC107219.2 | AAI07220.1 | |||
| BC134353.1 |
More Information
Set Membership: |
|

Genomic Map