Assay Details
Target Gene Details
Entrez Gene ID: | 16331 |
Gene Name: | inositol polyphosphate-5-phosphatase D |
Gene Aliases: |
SHIP, SHIP-1, SHIP1, SIP-145, p150Ship, s-SHIP |
Location: |
Chr.1:87620312-87720510 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inpp5d | NM_001110192.2 | NP_001103662.1 | ||
| NM_001110193.2 | NP_001103663.1 | |||
| NM_010566.3 | NP_034696.2 | |||
| NR_122078.1 | ||||
| AF125996.1 | AAD37118.1 | |||
| AF184912.1 | AAF25823.1 | |||
| AF184913.1 | AAF25824.1 | |||
| AF228679.1 | AAF69143.1 | |||
| AK143560.1 | BAE25436.1 | |||
| BC108328.1 | AAI08329.1 | |||
| BG144006.1 | ||||
| U39203.1 | AAB18937.1 | |||
| U51742.1 | AAC52606.1 | |||
| U52044.1 | AAC53023.1 |

Genomic Map