Assay Details
Target Gene Details
Entrez Gene ID: | 16978 |
Gene Name: | leucine rich repeat (in FLII) interacting protein 1 |
Gene Aliases: |
AU024550, Fliiap1 |
Location: |
Chr.1:90998458-91128944 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrfip1 | NM_001111311.1 | NP_001104781.1 | ||
| NM_001111312.1 | NP_001104782.1 | |||
| NM_008515.4 | NP_032541.1 | |||
| AF045573.1 | AAC40072.1 | |||
| AK041162.1 | BAC30844.2 | |||
| AK044174.1 | BAC31805.1 | |||
| AK077637.1 | ||||
| AK133661.1 | ||||
| AK134120.1 | BAE22022.1 | |||
| BC144956.1 | ||||
| BC145642.1 | ||||
| BY739242.1 | ||||
| CJ154753.1 |
More Information
Set Membership: |
|

Genomic Map