Assay Details
Target Gene Details
Entrez Gene ID: | 98682 |
Gene Name: | major facilitator superfamily domain containing 6 |
Gene Aliases: |
2210010L05Rik, 9630025I22Rik, AI836200, AW212394, MMR2 |
Location: |
Chr.1:52656299-52727462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd6 | NM_133829.2 | NP_598590.2 | ||
| NM_178081.4 | NP_835182.1 | |||
| AB247936.1 | ||||
| AK035995.1 | BAC29272.1 | |||
| AK046866.1 | BAC32900.1 | |||
| BC017534.1 | AAH17534.1 |

Genomic Map