Assay Details
Target Gene Details
Entrez Gene ID: | 19070 |
Gene Name: | MOB family member 4, phocein |
Gene Aliases: |
2610109B12Rik, Mobkl3, Phocn, Prei3 |
Location: |
Chr.1:55130936-55154899 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mob4 | NM_025283.3 | NP_079559.2 | ||
| AK020446.1 | BAB32105.1 | |||
| AK147857.1 | BAE28182.1 | |||
| AK162327.1 | BAE36855.1 | |||
| BC037499.1 | AAH37499.1 | |||
| BC058168.1 | AAH58168.1 | |||
| BY768556.1 | ||||
| U01138.1 |
More Information
Set Membership: |
|

Genomic Map