Assay Details
Target Gene Details
Entrez Gene ID: | 66495 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3 |
Gene Aliases: |
2700033I16Rik, AI415450 |
Location: |
Chr.1:58586599-58595948 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufb3 | NM_025597.2 | NP_079873.1 | ||
| AK007930.1 | BAB25357.1 | |||
| AK009008.1 | BAB26021.1 | |||
| AK012320.1 | BAB28160.1 | |||
| AK013608.1 | BAB28928.1 | |||
| AK138123.1 | BAE23555.1 | |||
| BC036989.1 | AAH36989.1 |
More Information
Set Membership: |
|

Genomic Map