Assay Details
Target Gene Details
Entrez Gene ID: | 65102 |
Gene Name: | Ngg1 interacting factor 3-like 1 (S. pombe) |
Gene Aliases: |
1110030G24Rik |
Location: |
Chr.1:58445156-58462276 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nif3l1 | AF284439.1 | AAG45961.1 | ||
| AK003978.1 | BAB23102.1 | |||
| AK011670.1 | BAB27769.1 | |||
| AK131631.1 | BAE20729.1 | |||
| AK154815.1 | BAE32848.1 | |||
| AK158616.1 | BAE34583.1 | |||
| BC089475.1 | AAH89475.1 |

Genomic Map