Assay Details
Target Gene Details
Entrez Gene ID: | 271697 |
Gene Name: | cyclin-dependent kinase 15 |
Gene Aliases: |
Als2cr7, Pftk2 |
Location: |
Chr.1:59256889-59352612 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk15 | NM_001033373.2 | NP_001028545.2 | ||
| BC147314.1 | ||||
| BC147315.1 | ||||
| BC147758.1 | ||||
| BC147763.1 | ||||
| BC172036.1 |
More Information
Set Membership: |
|

Genomic Map