Assay Details
Target Gene Details
Entrez Gene ID: | 17901 |
Gene Name: | myosin, light polypeptide 1 |
Gene Aliases: |
AI325107, MLC1f, MLC3f, Mylf |
Location: |
Chr.1:66924295-66945473 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myl1 | NM_001113387.1 | NP_001106858.1 | ||
| NM_021285.3 | NP_067260.1 | |||
| AK003182.1 | BAB22625.1 | |||
| AK009938.1 | BAB26597.1 | |||
| BC059087.1 | AAH59087.1 | |||
| BY785713.1 | ||||
| W87050.1 |
More Information
Set Membership: |
|

Genomic Map