Assay Details
Target Gene Details
Entrez Gene ID: | 70225 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 3 |
Gene Aliases: |
2310076N22Rik, 2510026K04Rik, Cyp10l |
Location: |
Chr.1:58430993-58445527 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppil3 | NM_001285826.1 | NP_001272755.1 | ||
| NM_001285827.1 | NP_001272756.1 | |||
| NM_027351.3 | NP_081627.1 | |||
| NM_027374.3 | NP_081650.2 | |||
| AF315803.1 | AAO32944.1 | |||
| AK010207.1 | BAB26768.1 | |||
| AK011000.1 | BAB27319.1 | |||
| BB844274.1 | ||||
| BC060993.1 | AAH60993.1 | |||
| BC061645.1 | AAH61645.1 | |||
| BY288296.1 | ||||
| CJ166559.1 | ||||
| CJ170065.1 | ||||
| DV043111.1 |
More Information
Set Membership: |
|

Genomic Map