Assay Details
Target Gene Details
Entrez Gene ID: | 226407 |
Gene Name: | RAB3 GTPase activating protein subunit 1 |
Gene Aliases: |
1700003B17Rik, 4732493F09Rik, AL117896, Rab3gap, p130 |
Location: |
Chr.1:127868737-127943876 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab3gap1 | NM_178690.4 | NP_848805.2 | ||
| AK076244.1 | BAC36271.1 | |||
| AK076399.1 | BAC36323.1 | |||
| AK080432.1 | BAC37915.1 | |||
| AK134604.1 | BAE22204.1 | |||
| AK164523.1 | BAE37821.1 | |||
| AK170768.1 | ||||
| AK172881.1 | BAD32159.1 | |||
| BC046297.1 | AAH46297.1 |
More Information
Set Membership: |
|

Genomic Map