Assay Details
Target Gene Details
Entrez Gene ID: | 76816 |
Gene Name: | serologically defined colon cancer antigen 8 |
Gene Aliases: |
2700048G21Rik, 5730470G24Rik, Cccap |
Location: |
Chr.1:176814526-177020437 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdccag8 | NM_029756.3 | NP_084032.1 | ||
| AK012392.1 | BAB28209.1 | |||
| AK017691.1 | BAB30877.1 | |||
| AK051683.1 | BAC34719.1 | |||
| AK132316.1 | BAE21100.1 | |||
| AK133622.1 | BAE21753.1 | |||
| AK136921.1 | BAE23170.1 | |||
| BC016244.1 | ||||
| BC025138.1 | AAH25138.1 |

Genomic Map