Assay Details
Target Gene Details
Entrez Gene ID: | 66052 |
Gene Name: | succinate dehydrogenase complex, subunit C, integral membrane protein |
Gene Aliases: |
0610010E03Rik, AI316496, AU019277 |
Location: |
Chr.1:171129157-171150603 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdhc | NM_025321.3 | NP_079597.2 | ||
| AK002459.1 | BAB22116.1 | |||
| AK012818.1 | BAB28491.1 | |||
| AK135503.1 | BAE22557.1 | |||
| BC005779.1 | AAH05779.1 | |||
| BC083091.1 | AAH83091.1 | |||
| BY711932.1 |
More Information
Set Membership: |
|

Genomic Map